missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99+%

CAS: 872-50-4 | C5H9NO | 99.13 g/mol

Supplier:  Thermo Scientific Chemicals A122600E

 View more versions of this product

Catalog No. AAA122600E

Only null left
Explore available promotions

Description

Description

1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.

Solubility
Miscible with water, lower alcohols, ketones, ethyl acetate, castor oil, chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents. Hygroscopic.
Specifications

Specifications

1-Methyl-2-pyrrolidinone
872-50-4
204°C
86°C (187°F)
1.4705
MFCD00003193
Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
CN1CCCC1=O
99.13
-24°C
1.033
2500 g
Amine-like
C5H9NO
106420
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
99.13

Chemical Identifiers

872-50-4
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
CN1CCCC1=O
C5H9NO
MFCD00003193
1-methylpyrrolidin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335-H360D-H373

EINECSNumber : 212-828-1

RTECSNumber : UY5790000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only