missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy

CAS: 872-50-4 | C5H9NO | 99.133 g/mol

Supplier:  Thermo Scientific Chemicals 222080025

 View more versions of this product

Catalog No. AC222080025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For UV/VIS spectroscopy
Specifications

Specifications

1-Methyl-2-pyrrolidinone
872-50-4
1.0300g/mL
Glass Bottle
99+%
MFCD00003193
0.05% max. (K.F.)
01,696; 02,281; 09,316; 11,346
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
CN1CCCC1=O
99.133
CHEBI:7307
≥99.0%
99+%
(1 cm cell vs water), 0.01 at 340nm, 0.04 at 320nm, 0.1 at 300nm, 0.55 at 270nm, 1 at 262nm
2.5 L
Liquid
C5H9NO
Vapor Pressure: 0.7mbar at 25°C
21, 237
15, 6190
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
13387
99.13
Spectroscopy

Chemical Identifiers

872-50-4
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
13387
1-methylpyrrolidin-2-one
C5H9NO
MFCD00003193
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
CHEBI:7307
CN1CCCC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 212-828-1

RTECSNumber : UY5790000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only