missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

Catalog No. p-216815
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC364381000 100 mL AcroSeal™ Glass Bottle
AC364380010 1 L AcroSeal™ Glass Bottle
AC364380025 2.5 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC364381000 Supplier Thermo Scientific Chemicals Supplier No. 364381000
Only null left

CAS: 872-50-4 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24.0°C
Color Colorless
CAS Min % 99.4
CAS Max % 100.0
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 100 mL
Flash Point 91°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.5%
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Beilstein 21, 237
Water 0.005% Max. (Coulometric)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material 1-Methyl-2-pyrrolidinone, Over Molecular Sieves
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-828-1
RTECSNumber UY5790000
TSCA TSCA

RUO – Research Use Only