missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 98%, pure

Catalog No. p-4812625
Change view
Click to view available options
Quantity:
5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449180050 5 L
AC449180250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449180050 Supplier Thermo Scientific Chemicals Supplier No. 449180050
Only null left

1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O | CAS: 872-50-4 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24.0°C
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 5 L
Flash Point 91°C
Packaging Metal Drum
Refractive Index 1.4690 to 1.4710
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 98%
Grade Pure
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 212-828-1

RUO – Research Use Only