missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolecarboxylic Acid 98.0+%, TCI America™
$178.95 - $178.95
Chemical Identifiers
| CAS | 6973-60-0 |
|---|---|
| Molecular Formula | C6H7NO2 |
| Molecular Weight (g/mol) | 125.127 |
| MDL Number | MFCD00003088 |
| InChI Key | ILAOVOOZLVGAJF-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole |
| PubChem CID | 81453 |
| IUPAC Name | 1-methylpyrrole-2-carboxylic acid |
| SMILES | CN1C=CC=C1C(=O)O |
Chemical Identifiers
| 6973-60-0 | |
| 125.127 | |
| ILAOVOOZLVGAJF-UHFFFAOYSA-N | |
| 81453 | |
| CN1C=CC=C1C(=O)O |
| C6H7NO2 | |
| MFCD00003088 | |
| 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole | |
| 1-methylpyrrole-2-carboxylic acid |
Specifications
| 6973-60-0 | |
| C6H7NO2 | |
| 1 g | |
| ILAOVOOZLVGAJF-UHFFFAOYSA-N | |
| 1-methylpyrrole-2-carboxylic acid | |
| 81453 | |
| ≥98.0% (GC,T) | |
| 1-Methyl-2-pyrrolecarboxylic Acid |
| 136°C | |
| MFCD00003088 | |
| 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole | |
| CN1C=CC=C1C(=O)O | |
| 125.127 | |
| 125.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No