missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolecarboxylic Acid 98.0+%, TCI America™

Catalog No. M21611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21611G Supplier TCI America Supplier No. M21611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6973-60-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00003088
InChI Key ILAOVOOZLVGAJF-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole
PubChem CID 81453
IUPAC Name 1-methylpyrrole-2-carboxylic acid
SMILES CN1C=CC=C1C(=O)O

Specifications

Melting Point 136°C
Quantity 1 g
Formula Weight 125.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-2-pyrrolecarboxylic Acid