missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-(methylsulfonyl)benzimidazole 98.0+%, TCI America™

Catalog No. p-7133477
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28601G 1 g
M28605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28601G Supplier TCI America Supplier No. M28601G
Only null left

Chemical Identifiers

CAS 61078-14-6
Molecular Formula C9H10N2O2S
Molecular Weight (g/mol) 210.251
InChI Key VDZVTXFWEKBHBV-UHFFFAOYSA-N
PubChem CID 640715
IUPAC Name 1-methyl-2-methylsulfonylbenzimidazole
SMILES CN1C2=CC=CC=C2N=C1S(=O)(=O)C

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-2-(methylsulfonyl)benzimidazole