missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl 2-Aminoterephthalate 98.0+%, TCI America™

Catalog No. p-7117483
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15075G Supplier TCI America Supplier No. A15075G
Only null left

Chemical Identifiers

CAS 60728-41-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00189374
InChI Key QKOKLMFCKLEFDV-UHFFFAOYSA-N
Synonym 3-Amino-4-carbomethoxybenzoic Acid, 3-Amino-4-methoxycarbonylbenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester
PubChem CID 5235500
IUPAC Name 3-amino-4-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 218°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl 2-Aminoterephthalate