missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-(2-pyridyl)pyridinium Iodide 98.0+%, TCI America™

Catalog No. M2778200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2778200MG 200 mg
M27781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2778200MG Supplier TCI America Supplier No. M2778200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77972-47-5
Molecular Formula C11H12I2N2
Molecular Weight (g/mol) 426.04
InChI Key VKILBCPXPJNJFR-UHFFFAOYSA-M
PubChem CID 71403174
IUPAC Name 1-methyl-2-pyridin-1-ium-2-ylpyridin-1-ium;diiodide
SMILES C[N+]1=CC=CC=C1C2=CC=CC=[NH+]2.[I-].[I-]

Specifications

Melting Point 149°C
Quantity 200 mg
Formula Weight 298.13
Percent Purity ≥98.0% (HPLC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-2-(2-pyridyl)pyridinium Iodide