accessibility menu, dialog, popup

UserName

CH3NNI

CAS RN 77972-47-5

IUPAC Name: 1-methyl-[2,2'-bipyridin]-1-ium iodide
Synonyms: 1-methyl-[2,2'-bipyridin]-1-ium iodide
Molecular Weight (g/mol): 298.13
Molecular Formula: C11H11IN2
InChi Key: PIKLCVQDBVTHFG-UHFFFAOYSA-M
SMILES: [I-].C[N+]1=CC=CC=C1C1=CC=CC=N1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results