missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-carboxaldehyde, 96%

Catalog No. AAH5414603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5414603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5414603 Supplier Thermo Scientific Chemicals Supplier No. H5414603

CAS: 25016-11-9 | C5H6N2O | 110.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25016-11-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00460465
InChI Key MYFZXSOYJVWTBL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carbaldehyde, 1-methyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1-methyl, 4-formyl-1-methylpyrazole, 4-formyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-carboxaldehyde, 1-methylpyrazole-4-carboxyaldehyde, 1-methyl-1h-pyrazol-4-carbaldehyd, 1-methylpyrazole-4-aldehyde, acmc-1cs06
PubChem CID 573117
IUPAC Name 1-methylpyrazole-4-carbaldehyde
SMILES CN1C=C(C=N1)C=O

Specifications

Melting Point 29°C to 34°C
Flash Point >230°C (446°F)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 110.12
Percent Purity 96%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-carboxaldehyde
Hazard Category H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only