missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC77402DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC77402DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

79583-19-0
144.558
QLBBQLJPRXPVOS-UHFFFAOYSA-N
3164931
CN1C=C(C=N1)C(=O)Cl
C5H5ClN2O
MFCD03421496
1-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl, 1h-pyrazole-4-carbonylchloride, 1-methyl, 1-methyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-9ci
1-methylpyrazole-4-carbonyl chloride

Specifications

1-Methyl-1H-pyrazole-4-carbonyl chloride
96.5
97%
C5H5ClN2O
MFCD03421496
QLBBQLJPRXPVOS-UHFFFAOYSA-N
1-methylpyrazole-4-carbonyl chloride
3164931
97%
79583-19-0
100.0
Amber Glass Bottle
1g
1-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl, 1h-pyrazole-4-carbonylchloride, 1-methyl, 1-methyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-9ci
CN1C=C(C=N1)C(=O)Cl
144.558
144.56
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents