CAS RN 79583-19-0
IUPAC Name:
1-methyl-1H-pyrazole-4-carbonyl chloride
Synonyms:
1-methylpyrazole-4-carbonyl chloride
Molecular Weight (g/mol):
144.56
Molecular Formula:
C5H5ClN2O
InChi Key:
QLBBQLJPRXPVOS-UHFFFAOYSA-N
SMILES:
CN1C=C(C=N1)C(Cl)=O
1
–
1
of
1
results
1
1-methyl-1h-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™
CAS: 79583-19-0 Molecular Formula: C5H5ClN2O Molecular Weight (g/mol): 144.558 MDL Number: MFCD03421496 InChI Key: QLBBQLJPRXPVOS-UHFFFAOYSA-N Synonym: 1-methyl-1h-pyrazole-4-carbonyl chloride,1h-pyrazole-4-carbonyl chloride, 1-methyl,1h-pyrazole-4-carbonylchloride, 1-methyl,1-methyl-1h-pyrazole-4-carboxylic acid chloride,1h-pyrazole-4-carbonyl chloride, 1-methyl-9ci PubChem CID: 3164931 IUPAC Name: 1-methylpyrazole-4-carbonyl chloride SMILES: CN1C=C(C=N1)C(=O)Cl