missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC77402CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC77402CB 250mg Amber Glass Bottle
CC77402DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC77402CB Supplier Thermo Scientific Chemicals Supplier No. CC77402CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79583-19-0
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD03421496
InChI Key QLBBQLJPRXPVOS-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl, 1h-pyrazole-4-carbonylchloride, 1-methyl, 1-methyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-9ci
PubChem CID 3164931
IUPAC Name 1-methylpyrazole-4-carbonyl chloride
SMILES CN1C=C(C=N1)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.