missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-boronic acid hydrochloride, 95%, Thermo Scientific™

Chemical Identifiers

CAS 1026796-02-0
Molecular Formula C4H7BN2O2·ClH
Molecular Weight (g/mol) 162.38
InChI Key YFPLZINTZVVVJM-UHFFFAOYSA-N
Synonym 1-methylpyrazole-4-boronic acid, hcl, 1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid,hcl, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl
PubChem CID 45158882
IUPAC Name (1-methylpyrazol-4-yl)boronic acid;hydrochloride
SMILES B(C1=CN(N=C1)C)(O)O.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC445490010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1026796-02-0
162.38
1-methylpyrazole-4-boronic acid, hcl, 1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid,hcl, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl
(1-methylpyrazol-4-yl)boronic acid;hydrochloride
C4H7BN2O2·ClH
YFPLZINTZVVVJM-UHFFFAOYSA-N
45158882
B(C1=CN(N=C1)C)(O)O.Cl

Specifications

1026796-02-0
Glass Bottle
1-methylpyrazole-4-boronic acid, hcl, 1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid,hcl, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl
B(C1=CN(N=C1)C)(O)O.Cl
162.38
162.38
1-Methyl-1H-pyrazole-4-boronic acid hydrochloride
94% min. (HPLC)
C4H7BN2O2·ClH
YFPLZINTZVVVJM-UHFFFAOYSA-N
(1-methylpyrazol-4-yl)boronic acid;hydrochloride
45158882
95%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only