accessibility menu, dialog, popup

UserName

BNNCH3HOOHHCl

CAS RN 1026796-02-0

IUPAC Name: (1-methyl-1H-pyrazol-4-yl)boronic acid hydrochloride
Synonyms: 1-methylpyrazol-4-ylboronic acid hydrochloride
Molecular Weight (g/mol): 162.38
Molecular Formula: C4H8BClN2O2
InChi Key: YFPLZINTZVVVJM-UHFFFAOYSA-N
SMILES: Cl.CN1C=C(C=N1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)