missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazol-3-amine, 97%

Catalog No. AC456730050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456730050 5 g
AC456730250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456730050 Supplier Thermo Scientific Chemicals Supplier No. 456730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1904-31-0 | C4H7N3 | 97.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1904-31-0
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.12
InChI Key MOGQNVSKBCVIPW-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-3-amine, 3-amino-1-methylpyrazole, 1-methyl-3-aminopyrazole, 3-amino-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazol-3-ylamine, 3-amino-1-methyl pyrazole, 1h-pyrazol-3-amine, 1-methyl, 1-methylpyrazole-3-ylamine, 3-amino-1-methyl-pyrazole, 3-amino-1 methyl pyrazole
PubChem CID 137254
IUPAC Name 1-methylpyrazol-3-amine
SMILES CN1C=CC(=N1)N

Specifications

Boiling Point 93.0°C to 94.0°C (0.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazol-3-amine
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

RUO – Research Use Only