missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-indole-7-sulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66903CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC66903CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

941716-95-6
229.678
DVEZDWQOMVECCI-UHFFFAOYSA-N
24229740
CN1C=CC2=C1C(=CC=C2)S(=O)(=O)Cl
C9H8ClNO2S
MFCD09879958
1-methyl-1h-indole-7-sulfonyl chloride, 1-methyl-1h-indole-7-sulphonyl chloride, 1h-indole-7-sulfonylchloride, 1-methyl, 1h-indole-7-sulfonyl chloride, 1-methyl, chloro 1-methylindol-7-yl sulfone, 7-chlorosulphonyl-1-methyl-1h-indole
1-methylindole-7-sulfonyl chloride

Specifications

1-Methyl-1H-indole-7-sulfonyl chloride
96.5
97%
C9H8ClNO2S
MFCD09879958
DVEZDWQOMVECCI-UHFFFAOYSA-N
1-methylindole-7-sulfonyl chloride
24229740
97%
941716-95-6
100.0
Amber Glass Bottle
250mg
1-methyl-1h-indole-7-sulfonyl chloride, 1-methyl-1h-indole-7-sulphonyl chloride, 1h-indole-7-sulfonylchloride, 1-methyl, 1h-indole-7-sulfonyl chloride, 1-methyl, chloro 1-methylindol-7-yl sulfone, 7-chlorosulphonyl-1-methyl-1h-indole
CN1C=CC2=C1C(=CC=C2)S(=O)(=O)Cl
229.678
229.69
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions