CAS RN 941716-95-6
CAS RN 941716-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-methyl-1h-indole-7-sulfonyl chloride, 97%, Thermo Scientific™
CAS: 941716-95-6 Molecular Formula: C9H8ClNO2S Molecular Weight (g/mol): 229.678 MDL Number: MFCD09879958 InChI Key: DVEZDWQOMVECCI-UHFFFAOYSA-N Synonym: 1-methyl-1h-indole-7-sulfonyl chloride,1-methyl-1h-indole-7-sulphonyl chloride,1h-indole-7-sulfonylchloride, 1-methyl,1h-indole-7-sulfonyl chloride, 1-methyl,chloro 1-methylindol-7-yl sulfone,7-chlorosulphonyl-1-methyl-1h-indole PubChem CID: 24229740 IUPAC Name: 1-methylindole-7-sulfonyl chloride SMILES: CN1C=CC2=C1C(=CC=C2)S(=O)(=O)Cl