missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-3-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC06304DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC06304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

4002-83-9
160.18
KYDLGYCLCFKUHY-UHFFFAOYSA-N
7164545
CN1N=C(C=O)C2=CC=CC=C12
C9H8N2O
MFCD08060478
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
1-methylindazole-3-carbaldehyde

Specifications

1-Methyl-1H-indazole-3-carbaldehyde
96.5
97%
C9H8N2O
MFCD08060478
KYDLGYCLCFKUHY-UHFFFAOYSA-N
1-methylindazole-3-carbaldehyde
7164545
97%
4002-83-9
100.0
Amber Glass Bottle
1g
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
CN1N=C(C=O)C2=CC=CC=C12
160.18
160.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents