missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-5-carbonyl chloride hydrochloride, Thermo Scientific™

Catalog No. CC23602DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23602DA 1 g Amber Glass Bottle
CC23602CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC23602DA Supplier Thermo Scientific Chemicals Supplier No. CC23602DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 343569-06-2
Molecular Formula C5H6Cl2N2O
Molecular Weight (g/mol) 181.02
MDL Number MFCD03659705
InChI Key IRADJQMSOFSMNS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-5-carbonyl chloride hydrochloride, 3-methylimidazole-4-carbonyl chloride hydrochloride, 1-methylimidazole-5-carbonyl chloride hcl, 3-methyl-3h-imidazole-4-carbonyl chloride hydrochloride, 3-methyl-4-imidazolecarbonyl chloride hydrochloride, 5-chlorocarbonyl-1-methyl-1h-imidazole hydrochloride, 1h-imidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
PubChem CID 2776553
SMILES Cl.CN1C=NC=C1C(Cl)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 3-methylimidazole-4-carbonyl chloride;hydrochloride
Formula Weight 181.02
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonyl chloride hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.