accessibility menu, dialog, popup

UserName

CAS RN 343569-06-2

IUPAC Name: 1-methyl-1H-imidazole-5-carbonyl chloride hydrochloride
Synonyms: 3-methylimidazole-4-carbonyl chloride hydrochloride
Molecular Weight (g/mol): 181.02
Molecular Formula: C5H6Cl2N2O
InChi Key: IRADJQMSOFSMNS-UHFFFAOYSA-N
SMILES: Cl.CN1C=NC=C1C(Cl)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results