missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4,5-dicarboxylic Acid 97.0+%, TCI America™

Catalog No. M21565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21565G 5 g
M21561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21565G Supplier TCI America Supplier No. M21565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19485-38-2
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD01552045
InChI Key YARDQACXPOQDMO-UHFFFAOYSA-L
Synonym 4,5-Dicarboxy-1-methyl-1H-imidazole
PubChem CID 152258
IUPAC Name 1-methyl-1H-imidazole-4,5-dicarboxylate
SMILES CN1C=NC(C([O-])=O)=C1C([O-])=O

Specifications

Melting Point 262°C
Color Yellow
Quantity 5 g
Formula Weight 170.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1H-imidazole-4,5-dicarboxylic Acid