accessibility menu, dialog, popup

UserName

CH3NNOOOO

CAS RN 19485-38-2

IUPAC Name: 1-methyl-1H-imidazole-4,5-dicarboxylate
Synonyms: 1-methylimidazole-4,5-dicarboxylate
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: YARDQACXPOQDMO-UHFFFAOYSA-L
SMILES: CN1C=NC(C([O-])=O)=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results