missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 53484-17-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD06809665
InChI Key KAJLCBKTWRUQOI-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
PubChem CID 4913063
IUPAC Name 1-methylbenzimidazole-5-carboxylic acid
SMILES CN1C=NC2=C1C=CC(=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC13601CB
SDS View Documents
Thermo Scientific Chemicals
CC13601CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC13601DA
SDS View Documents
Thermo Scientific Chemicals
CC13601DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

53484-17-6
176.175
KAJLCBKTWRUQOI-UHFFFAOYSA-N
4913063
CN1C=NC2=C1C=CC(=C2)C(=O)O
C9H8N2O2
MFCD06809665
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
1-methylbenzimidazole-5-carboxylic acid

Specifications

53484-17-6
100.0
Amber Glass Bottle
MFCD06809665
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
CN1C=NC2=C1C=CC(=C2)C(=O)O
176.175
176.17
1-Methyl-1H-benzimidazole-5-carboxylic acid
96.5
97%
C9H8N2O2
250mg
KAJLCBKTWRUQOI-UHFFFAOYSA-N
1-methylbenzimidazole-5-carboxylic acid
4913063
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'