missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC13601DA

 View more versions of this product

Catalog No. CC13601DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-Methyl-1H-benzimidazole-5-carboxylic acid
96.5
97%
C9H8N2O2
1g
KAJLCBKTWRUQOI-UHFFFAOYSA-N
1-methylbenzimidazole-5-carboxylic acid
4913063
97%
53484-17-6
100.0
Amber Glass Bottle
MFCD06809665
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
CN1C=NC2=C1C=CC(=C2)C(=O)O
176.175
176.17

Chemical Identifiers

53484-17-6
176.175
KAJLCBKTWRUQOI-UHFFFAOYSA-N
4913063
CN1C=NC2=C1C=CC(=C2)C(=O)O
C9H8N2O2
MFCD06809665
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
1-methylbenzimidazole-5-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions