missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC23504DA

Quantity 1g

 View more versions of this product

Catalog No. CC23504DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

499770-67-1
161.164
1-methyl-1h-benzo d 1,2,3 triazole-5-carbaldehyde, 1-methyl-1h-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-1h-benzotriazole-5-carbaldehyde, 1h-benzotriazole-5-carboxaldehyde, 1-methyl, 1-methyl-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-5-benzotriazolecarboxaldehyde, 1-methylbenzotriazole-5-carboxaldehyde
1-methylbenzotriazole-5-carbaldehyde
C8H7N3O
IPVPVOVAJDRRAK-UHFFFAOYSA-N
2795417
CN1C2=C(C=C(C=C2)C=O)N=N1

Specifications

1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde
97%
C8H7N3O
1-methyl-1h-benzo d 1,2,3 triazole-5-carbaldehyde, 1-methyl-1h-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-1h-benzotriazole-5-carbaldehyde, 1h-benzotriazole-5-carboxaldehyde, 1-methyl, 1-methyl-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-5-benzotriazolecarboxaldehyde, 1-methylbenzotriazole-5-carboxaldehyde
CN1C2=C(C=C(C=C2)C=O)N=N1
161.164
161.16
499770-67-1
Amber Glass Bottle
1g
IPVPVOVAJDRRAK-UHFFFAOYSA-N
1-methylbenzotriazole-5-carbaldehyde
2795417
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions