missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog No. p-4510253
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC23504CB 250mg
CC23504DA 1g
CC23504DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC23504CB Supplier Thermo Scientific Chemicals Supplier No. CC23504CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 499770-67-1
Molecular Formula C8H7N3O
Molecular Weight (g/mol) 161.164
InChI Key IPVPVOVAJDRRAK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzo d 1,2,3 triazole-5-carbaldehyde, 1-methyl-1h-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-1h-benzotriazole-5-carbaldehyde, 1h-benzotriazole-5-carboxaldehyde, 1-methyl, 1-methyl-1,2,3-benzotriazole-5-carbaldehyde, 1-methyl-5-benzotriazolecarboxaldehyde, 1-methylbenzotriazole-5-carboxaldehyde
PubChem CID 2795417
IUPAC Name 1-methylbenzotriazole-5-carbaldehyde
SMILES CN1C2=C(C=C(C=C2)C=O)N=N1

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde