missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-propylpyrrolidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$182.68 - $542.44
Chemical Identifiers
| CAS | 223437-05-6 |
|---|---|
| Molecular Formula | C10H18F6N2O4S2 |
| Molecular Weight (g/mol) | 408.374 |
| MDL Number | MFCD08458917 |
| InChI Key | DKNRELLLVOYIIB-UHFFFAOYSA-N |
| PubChem CID | 25171607 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium |
| SMILES | CCC[N+]1(CCCC1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 223437-05-6 | |
| 408.374 | |
| DKNRELLLVOYIIB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium |
| C10H18F6N2O4S2 | |
| MFCD08458917 | |
| 25171607 | |
| CCC[N+]1(CCCC1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Specifications
| 223437-05-6 | |
| Yellow | |
| MFCD08458917 | |
| DKNRELLLVOYIIB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium | |
| 25171607 | |
| ≥98.0% (T) | |
| 1-Methyl-1-propylpyrrolidinium Bis(trifluoromethanesulfonyl)imide |
| 12°C | |
| C10H18F6N2O4S2 | |
| 5 g | |
| CCC[N+]1(CCCC1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 408.374 | |
| 408.37 | |
| Liquid |
Safety and Handling
TSCA : No