missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-propylpyrrolidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
Supplier: TCI America M20985G
Specifications
| 1-Methyl-1-propylpyrrolidinium Bis(trifluoromethanesulfonyl)imide | |
| 12°C | |
| C10H18F6N2O4S2 | |
| 5 g | |
| CCC[N+]1(CCCC1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 408.374 | |
| 408.37 | |
| Liquid |
| 223437-05-6 | |
| Yellow | |
| MFCD08458917 | |
| DKNRELLLVOYIIB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium | |
| 25171607 | |
| ≥98.0% (T) |
Chemical Identifiers
| 223437-05-6 | |
| 408.374 | |
| DKNRELLLVOYIIB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-1-propylpyrrolidin-1-ium |
| C10H18F6N2O4S2 | |
| MFCD08458917 | |
| 25171607 | |
| CCC[N+]1(CCCC1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Safety and Handling
TSCA : No