missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-propylpiperidinium Bromide 98.0+%, TCI America™
Supplier: TCI America M197625G
Specifications
| 1-Methyl-1-propylpiperidinium Bromide | |
| 248°C | |
| C9H20BrN | |
| 25 g | |
| [Br-].CCC[N+]1(C)CCCCC1 | |
| 222.17 | |
| 222.17 | |
| Crystalline Powder |
| 88840-42-0 | |
| White-Yellow | |
| MFCD17014743 | |
| CSANMXKHSHGXNC-UHFFFAOYSA-M | |
| 1-methyl-1-propylpiperidin-1-ium bromide | |
| 60196385 | |
| ≥98.0% (T) |
Chemical Identifiers
| 88840-42-0 | |
| 222.17 | |
| CSANMXKHSHGXNC-UHFFFAOYSA-M | |
| 1-methyl-1-propylpiperidin-1-ium bromide |
| C9H20BrN | |
| MFCD17014743 | |
| 60196385 | |
| [Br-].CCC[N+]1(C)CCCCC1 |
Safety and Handling
TSCA : No