missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-propylpiperidinium Bromide 98.0+%, TCI America™

Catalog No. p-7132947
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19765G 5 g
M197625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19765G Supplier TCI America Supplier No. M19765G

Chemical Identifiers

CAS 88840-42-0
Molecular Formula C9H20BrN
Molecular Weight (g/mol) 222.17
MDL Number MFCD17014743
InChI Key CSANMXKHSHGXNC-UHFFFAOYSA-M
PubChem CID 60196385
IUPAC Name 1-methyl-1-propylpiperidin-1-ium bromide
SMILES [Br-].CCC[N+]1(C)CCCCC1

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1-propylpiperidinium Bromide