missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclopentene 98.0+%, TCI America™

Catalog No. p-7132324
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07275ML 5 mL
M072725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07275ML Supplier TCI America Supplier No. M07275ML
Only null left

Chemical Identifiers

CAS 693-89-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00001397
InChI Key ATQUFXWBVZUTKO-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
PubChem CID 12746
IUPAC Name 1-methylcyclopent-1-ene
SMILES CC1=CCCC1

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-cyclopentene