missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene, 96%

Catalog No. aal1501414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1501414 25 g
AAL1501406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1501414 Supplier Thermo Scientific Chemicals Supplier No. L1501414
Only null left

CAS: 591-49-1 | C7H12 | 96.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-49-1
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00001548
InChI Key CTMHWPIWNRWQEG-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
PubChem CID 11574
IUPAC Name 1-methylcyclohexene
SMILES CC1=CCCCC1

Specifications

Density 0.81
Boiling Point 108°C to 112°C
Flash Point −1°C (30°F)
Refractive Index 1.45
Quantity 25 g
UN Number UN3295
Beilstein 1304483
Formula Weight 96.17
Percent Purity 96%
Chemical Name or Material 1-Methyl-1-cyclohexene
Hazard Category H225-H304-H315-H320-H335
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P302+P352-P305+P351+P338-P312-P331-P332+P313-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-718-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only