missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-cyclohexanecarboxylic Acid 99.0+%, TCI America™
Supplier: TCI America M073225G
Specifications
| 1-Methyl-1-cyclohexanecarboxylic Acid | |
| 38°C | |
| 236°C | |
| MFCD00001463 | |
| 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference | |
| CC1(CCCCC1)C(=O)O | |
| 142.198 | |
| 142.20 | |
| Crystalline Powder |
| 1123-25-7 | |
| White | |
| C8H14O2 | |
| 25 g | |
| REHQLKUNRPCYEW-UHFFFAOYSA-N | |
| 1-methylcyclohexane-1-carboxylic acid | |
| 70744 | |
| ≥99.0% (GC,T) |
Chemical Identifiers
| 1123-25-7 | |
| 142.198 | |
| REHQLKUNRPCYEW-UHFFFAOYSA-N | |
| 70744 | |
| CC1(CCCCC1)C(=O)O |
| C8H14O2 | |
| MFCD00001463 | |
| 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference | |
| 1-methylcyclohexane-1-carboxylic acid |
Safety and Handling
RTECSNumber : GU8585500
TSCA : Yes