missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexanecarboxylic Acid 99.0+%, TCI America™

Catalog No. M07325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07325G 5 g
M073225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07325G Supplier TCI America Supplier No. M07325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1123-25-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00001463
InChI Key REHQLKUNRPCYEW-UHFFFAOYSA-N
Synonym 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference
PubChem CID 70744
IUPAC Name 1-methylcyclohexane-1-carboxylic acid
SMILES CC1(CCCCC1)C(=O)O

Specifications

Melting Point 38°C
Color White
Boiling Point 236°C
Quantity 5 g
Formula Weight 142.20
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-1-cyclohexanecarboxylic Acid
RTECSNumber GU8585500
TSCA Yes