missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,4-cyclohexadiene 95.0+%, TCI America™

Catalog No. M04865ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04865ML 5 mL
M048625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04865ML Supplier TCI America Supplier No. M04865ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4313-57-9
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00001538
InChI Key QDXQAOGNBCOEQX-UHFFFAOYSA-N
Synonym 1-methyl-1,4-cyclohexadiene, 2,5-dihydrotoluene, 1,4-cyclohexadiene, 1-methyl, 1, 1-methyl, acmc-1ast4, 1-methyl1,4-cyclohexadiene, 1-methylcyclohexane-1,4-diene, 1-methyl-1,4-cyclohexadiene, stabilized
PubChem CID 78006
IUPAC Name 1-methylcyclohexa-1,4-diene
SMILES CC1=CCC=CC1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 mL
UN Number 3295
Formula Weight 94.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1,4-cyclohexadiene