accessibility menu, dialog, popup

UserName

CAS RN 4313-57-9

IUPAC Name: 1-methylcyclohexa-1,4-diene
Synonyms: 1-methylcyclohexa-1,4-diene
Molecular Weight (g/mol): 94.16
Molecular Formula: C7H10
InChi Key: QDXQAOGNBCOEQX-UHFFFAOYSA-N
SMILES: CC1=CCC=CC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)