missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methoxypyrene 96.0+%, TCI America™
$146.31 - $442.79
Chemical Identifiers
| CAS | 34246-96-3 |
|---|---|
| Molecular Formula | C17H12O |
| Molecular Weight (g/mol) | 232.28 |
| MDL Number | MFCD00127747 |
| InChI Key | CPPFVJLHDNOVTN-UHFFFAOYSA-N |
| PubChem CID | 160236 |
| ChEBI | CHEBI:81610 |
| IUPAC Name | 1-methoxypyrene |
| SMILES | COC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
Chemical Identifiers
| 34246-96-3 | |
| 232.28 | |
| CPPFVJLHDNOVTN-UHFFFAOYSA-N | |
| CHEBI:81610 | |
| COC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 |
| C17H12O | |
| MFCD00127747 | |
| 160236 | |
| 1-methoxypyrene |
Specifications
| 34246-96-3 | |
| Yellow | |
| MFCD00127747 | |
| CPPFVJLHDNOVTN-UHFFFAOYSA-N | |
| 1-methoxypyrene | |
| 160236 | |
| 232.28 | |
| Crystalline Powder |
| 87°C | |
| C17H12O | |
| 1 g | |
| COC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34 | |
| 232.28 | |
| CHEBI:81610 | |
| ≥96.0% (GC) | |
| 1-Methoxypyrene |
Safety and Handling
TSCA : No