missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxypyrene 96.0+%, TCI America™

Catalog No. M29191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29191G 1 g
M29195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29191G Supplier TCI America Supplier No. M29191G
Only null left

Chemical Identifiers

CAS 34246-96-3
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.28
MDL Number MFCD00127747
InChI Key CPPFVJLHDNOVTN-UHFFFAOYSA-N
PubChem CID 160236
ChEBI CHEBI:81610
IUPAC Name 1-methoxypyrene
SMILES COC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 87°C
Color Yellow
Quantity 1 g
Formula Weight 232.28
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methoxypyrene