missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxynaphthalene 98.0+%, TCI America™

Catalog No. M0116500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0116500G 500 g
M011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0116500G Supplier TCI America Supplier No. M0116500G
Only null left

Chemical Identifiers

CAS 2216-69-5
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00003924
InChI Key NQMUGNMMFTYOHK-UHFFFAOYSA-N
Synonym naphthalene, 1-methoxy, methyl 1-naphthyl ether, 1-methoxy naphthalene, naphthalene, methoxy, alpha-methoxynaphthalene, unii-dg2eol57lf, .alpha.-methoxynaphthalene, methoxynaphthalene, alpha-naphthyl methyl ether, dg2eol57lf
PubChem CID 16668
IUPAC Name 1-methoxynaphthalene
SMILES COC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 6°C
Boiling Point 271°C
Quantity 500 g
UN Number 3082
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxynaphthalene
EINECSNumber (4)-0361
RTECSNumber QJ9465500
TSCA Yes