accessibility menu, dialog, popup

UserName

H3CO

CAS RN 2216-69-5

IUPAC Name: 1-methoxynaphthalene
Synonyms: naphthalene, 1-methoxy-
Molecular Weight (g/mol): 158.2
Molecular Formula: C11H10O
InChi Key: NQMUGNMMFTYOHK-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)