missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxynaphthalene, 98+%

Catalog No. p-7027800
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2482514 25 g
AAB2482522 100 g
AAB2482536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2482514 Supplier Thermo Scientific Chemicals Supplier No. B2482514

CAS: 2216-69-5 | C11H10O | 158.2 g/mol

1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methoxynaphthalene is used in the study of peroxygenase activity of cytochrome c peroxidase. It is used as a precursor to synthesize prenyl naphthalene-ols, which shows antioxidative activity.

Solubility
Immiscible with water. Miscible with ether, carbon disulfide and benzene.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2216-69-5
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00003924
InChI Key NQMUGNMMFTYOHK-UHFFFAOYSA-N
Synonym naphthalene, 1-methoxy, methyl 1-naphthyl ether, 1-methoxy naphthalene, naphthalene, methoxy, alpha-methoxynaphthalene, unii-dg2eol57lf, .alpha.-methoxynaphthalene, methoxynaphthalene, alpha-naphthyl methyl ether, dg2eol57lf
PubChem CID 16668
IUPAC Name 1-methoxynaphthalene
SMILES COC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 5°C
Density 1.094
Boiling Point 135°C to 137°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.621
Quantity 25 g
Beilstein 7748884
Solubility Information Immiscible with water. Miscible with ether,carbon disulfide and benzene.
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Methoxynaphthalene
Show More Show Less
EINECSNumber 218-696-1
RTECSNumber QJ9465500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only