missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-4-methylcyclohexane (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. M23755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23755G 5 g
M237525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23755G Supplier TCI America Supplier No. M23755G
Only null left

Chemical Identifiers

CAS 90200-72-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
InChI Key VBAPYXHNMYKBKN-UHFFFAOYSA-N
PubChem CID 6336264
IUPAC Name 1-methoxy-4-methylcyclohexane
SMILES CC1CCC(CC1)OC

Specifications

Color Colorless
Boiling Point 148°C
Quantity 5 g
UN Number 3271
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-4-methylcyclohexane (cis- and trans- mixture)