accessibility menu, dialog, popup

UserName

CAS RN 90200-72-9

CH3OCH3

CAS RN 90200-72-9

IUPAC Name: 1-methoxy-4-methylcyclohexane
Synonyms: 1-methoxy-4-methylcyclohexane
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: VBAPYXHNMYKBKN-UHFFFAOYSA-N
SMILES: COC1CCC(C)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results