missing translation for 'onlineSavingsMsg'
Learn More

1-Methanesulfonyl-2-imidazolidinone 98.0+%, TCI America™

Catalog No. M25535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25535G 5 g
M255325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25535G Supplier TCI America Supplier No. M25535G

Chemical Identifiers

CAS 41730-79-4
Molecular Formula C4H8N2O3S
Molecular Weight (g/mol) 164.179
MDL Number MFCD07787278
InChI Key WTCUHYRXLIHSLY-UHFFFAOYSA-N
PubChem CID 3016296
IUPAC Name 1-methylsulfonylimidazolidin-2-one
SMILES CS(=O)(=O)N1CCNC1=O

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 164.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methanesulfonyl-2-imidazolidinone