accessibility menu, dialog, popup

UserName

CH3SOONHNO

CAS RN 41730-79-4

IUPAC Name: 1-methanesulfonylimidazolidin-2-one
Synonyms: 1-methanesulfonylimidazolidin-2-one
Molecular Weight (g/mol): 164.18
Molecular Formula: C4H8N2O3S
InChi Key: WTCUHYRXLIHSLY-UHFFFAOYSA-N
SMILES: CS(=O)(=O)N1CCNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results