missing translation for 'onlineSavingsMsg'
Learn More

1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%

CAS: 74257-00-4 | C11H12N4O4S | 296.30 g/mol

$640.76 - $640.76

Chemical Identifiers

CAS 74257-00-4
Molecular Formula C11H12N4O4S
Molecular Weight (g/mol) 296.30
MDL Number MFCD00009754
InChI Key SFYDWLYPIXHPML-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-3-nitro-1h-1,2,4-triazole, 1-mesitylene-2-sulfonyl-3-nitro-1,2,4-triazole, msnt, 1-mesitylene-2-sulfonyl-3-nitro-1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole, 2,4,6-trimethylphenyl-3-nitro-1,2,4-triazol-1-yl sulfone, 3-nitro-1-2,4,6-trimethylbenzenesulfonyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl
PubChem CID 716901
IUPAC Name 3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221310050
SDS View Documents
Thermo Scientific Chemicals
221310050
5 g Glass bottle
Each for $640.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

74257-00-4
296.30
SFYDWLYPIXHPML-UHFFFAOYSA-N
716901
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
C11H12N4O4S
MFCD00009754
1-mesitylsulfonyl-3-nitro-1h-1,2,4-triazole, 1-mesitylene-2-sulfonyl-3-nitro-1,2,4-triazole, msnt, 1-mesitylene-2-sulfonyl-3-nitro-1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole, 2,4,6-trimethylphenyl-3-nitro-1,2,4-triazol-1-yl sulfone, 3-nitro-1-2,4,6-trimethylbenzenesulfonyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl
3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole

Specifications

74257-00-4
100.0
Yellow
97.5% min. (HPLC)
C11H12N4O4S
5 g
SFYDWLYPIXHPML-UHFFFAOYSA-N
3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
716901
98%
1-(Mesitylene-2-sulfonyl)-3-nitro-1, 2, 4-triazole, 98%
97.5
132.0°C to 134.0°C
Authentic
Glass bottle
MFCD00009754
1-mesitylsulfonyl-3-nitro-1h-1,2,4-triazole, 1-mesitylene-2-sulfonyl-3-nitro-1,2,4-triazole, msnt, 1-mesitylene-2-sulfonyl-3-nitro-1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole, 2,4,6-trimethylphenyl-3-nitro-1,2,4-triazol-1-yl sulfone, 3-nitro-1-2,4,6-trimethylbenzenesulfonyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
296.30
296.29
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only