missing translation for 'onlineSavingsMsg'
Learn More

1-(Mercaptomethyl)cyclopropaneacetic Acid 97.0+%, TCI America™

Catalog No. M20741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20741G 1 g
M20745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20741G Supplier TCI America Supplier No. M20741G

Chemical Identifiers

CAS 162515-68-6
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.204
MDL Number MFCD03411600
InChI Key VFAXPOVKNPTBTM-UHFFFAOYSA-N
Synonym 2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
PubChem CID 9793825
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
SMILES C1CC1(CC(=O)O)CS

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 146.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic Acid