accessibility menu, dialog, popup

UserName

OHOHS

CAS RN 162515-68-6

IUPAC Name: 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
Synonyms: [1-(sulfanylmethyl)cyclopropyl]acetic acid
Molecular Weight (g/mol): 146.2
Molecular Formula: C6H10O2S
InChi Key: VFAXPOVKNPTBTM-UHFFFAOYSA-N
SMILES: OC(=O)CC1(CS)CC1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)